siRNA Details
| Gene Symbol: Uggt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133596.1 | NP_598280.1 | 6 | 519 | ||
| XM_006244734.2 | XP_006244796.1 | 6 | 741 | |||
| XM_006244735.2 | XP_006244797.1 | 6 | 741 | |||
| GenBank mRNA | AF200359.1 | - | 6 | 519 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_198899, BC068283, AK087883, AK144381, AK050532, and others.

Uggt1
Genomic Map