siRNA Details
| Gene Symbol: Kcnh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133610.2 | NP_598294.1 | 4 | 634 | ||
| GenBank mRNA | AF185637.1 | - | 4 | 593 | ||
| AJ250280.1 | - | 4 | 636 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172805, AF309565, AK032438, BC066138.

Kcnh5
Genomic Map