siRNA Details
| Gene Symbol: Epha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134331.1 | NP_599158.1 | 3 | 425 | ||
| XM_006237962.2 | XP_006238024.1 | 3 | 583 | |||
| XM_006237963.2 | XP_006238025.1 | 3 | 583 | |||
| XM_006237964.2 | XP_006238026.1 | 3 | 580 | |||
| XM_006237965.2 | XP_006238027.1 | 3 | 581 | |||
| XM_008763587.1 | XP_008761809.1 | 3 | 579 | |||
| GenBank mRNA | U21954.1 | - | 3 | 425 | ||
| U21955.1 | - | 3 | 425 | |||

Epha7
Genomic Map