siRNA Details
| Gene Symbol: Folr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133527.1 | NP_598211.1 | 3 | 248 | ||
| GenBank mRNA | AF219904.1 | - | 3 | 248 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Folr1| Gene Symbol: Folr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133527.1 | NP_598211.1 | 3 | 248 | ||
| GenBank mRNA | AF219904.1 | - | 3 | 248 | ||