siRNA Details
| Gene Symbol: Pank4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133531.2 | NP_598215.1 | 12, 13 | 1587 | ||
| GenBank mRNA | AF399873.1 | - | 12, 13 | 1587 | ||
| FQ223969.1 | - | 12, 13 | 1594 | |||
| FQ224186.1 | - | 12, 13 | 1593 | |||

Pank4
Genomic Map