siRNA Details
| Gene Symbol: Ppp1r15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133546.3 | NP_598230.2 | 2 | 881 | ||
| XM_006228974.2 | XP_006229036.1 | 2 | 943 | |||
| GenBank mRNA | AF020618.1 | - | 1 | 1058 | ||
| BC072513.1 | - | 2 | 863 | 7104357 | ||

Ppp1r15a
Genomic Map