siRNA Details
Gene Symbol: Itfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133557.2 | NP_598241.2 | 2, 3 | 418 | ||
GenBank mRNA | AF480856.2 | - | 2, 3 | 294 | ||
BC085706.1 | - | 2, 3 | 418 | 7189102 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Itfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133557.2 | NP_598241.2 | 2, 3 | 418 | ||
GenBank mRNA | AF480856.2 | - | 2, 3 | 294 | ||
BC085706.1 | - | 2, 3 | 418 | 7189102 |