siRNA Details
| Gene Symbol: Lnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113403.1 | NP_001106874.1 | 3 | 1103 | ||
| NM_133574.1 | NP_598258.1 | 3 | 1103 | |||
| GenBank mRNA | U32990.1 | - | 3 | 1103 | ||
| U76997.1 | - | 3 | 1103 | |||
| Gene Symbol: LOC100912445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748706.3 | XP_003748754.1 | 5 | 1299 | ||
| XM_008758818.1 | XP_008757040.1 | 5 | 1299 | |||
| XM_008758819.1 | XP_008757041.1 | 5 | 1041 | |||

Lnpep
Genomic Map