siRNA Details
| Gene Symbol: Ndrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270862.1 | NP_001257791.1 | 9, 10 | 694 | ||
| NM_001270863.1 | NP_001257792.1 | 9, 10 | 676 | |||
| NM_001270864.1 | NP_001257793.1 | 8, 9 | 634 | |||
| NM_133583.2 | NP_598267.2 | 8, 9 | 652 | |||
| GenBank mRNA | AF334105.1 | - | 9, 10 | 658 | ||
| AF334106.1 | - | 8, 9 | 616 | |||
| AJ426424.1 | - | 9, 10 | 707 | |||
| AJ426425.1 | - | 8, 9 | 665 | |||
| AJ426426.1 | - | 9, 10 | 670 | |||
| AJ426427.1 | - | 8, 9 | 628 | |||
| FQ216465.1 | - | 1, 2 | 42 | |||

Ndrg2
Genomic Map