siRNA Details
| Gene Symbol: Opa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133585.3 | NP_598269.3 | 27 | 2874 | ||
| XM_006248497.1 | XP_006248559.1 | 29 | 3138 | |||
| XM_006248498.1 | XP_006248560.1 | 28 | 3084 | |||
| XM_006248499.1 | XP_006248561.1 | 28 | 3027 | |||
| GenBank mRNA | AY333988.1 | - | 27 | 2783 | ||
| AY510274.1 | - | 27 | 2929 | |||
| BC111071.1 | - | 27 | 2874 | 7324017 | ||
| U93197.1 | - | 11 | 1080 | |||

Opa1
Genomic Map