siRNA Details
| Gene Symbol: Uggt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133596.1 | NP_598280.1 | 21, 22 | 2225 | ||
| XM_006244734.2 | XP_006244796.1 | 21, 22 | 2447 | |||
| GenBank mRNA | AF200359.1 | - | 21, 22 | 2225 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uggt1| Gene Symbol: Uggt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133596.1 | NP_598280.1 | 21, 22 | 2225 | ||
| XM_006244734.2 | XP_006244796.1 | 21, 22 | 2447 | |||
| GenBank mRNA | AF200359.1 | - | 21, 22 | 2225 | ||