siRNA Details
| Gene Symbol: Clk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134340.1 | NP_599167.1 | 11, 12 | 1269 | ||
| XM_006243146.2 | XP_006243208.1 | 11, 12 | 2214 | |||
| XM_006243149.2 | XP_006243211.1 | 11, 12 | 1231 | |||
| XM_008766202.1 | XP_008764424.1 | 8, 9 | 963 | |||
| GenBank mRNA | BC070891.1 | - | 11, 12 | 1245 | 7099115 | |
| X94351.1 | - | 11, 12 | 1269 | |||

Clk3
Genomic Map