siRNA Details
| Gene Symbol: Trpm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134371.2 | NP_599198.2 | 3, 4 | 241 | ||
| XM_008767192.1 | XP_008765414.1 | 4, 5 | 503 | |||
| XM_008767193.1 | XP_008765415.1 | 4, 5 | 503 | |||
| GenBank mRNA | AY072788.2 | - | 3, 4 | 241 | ||

Trpm8
Genomic Map