siRNA Details
| Gene Symbol: Avpi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134373.2 | NP_599200.2 | 3 | 607 | ||
| XM_006231383.2 | XP_006231445.1 | 3 | 673 | |||
| GenBank mRNA | AJ294544.1 | - | 3 | 613 | ||
| FQ220140.1 | - | 3 | 517 | |||
| FQ220957.1 | - | 3 | 491 | |||

Avpi1
Genomic Map