siRNA Details
| Gene Symbol: Nlrp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134375.3 | NP_599202.2 | 1381 | |||
| GenBank mRNA | DQ631800.1 | - | 1381 | |||
| M85183.1 | - | 251 | ||||
| Gene Symbol: LOC103690037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230507.2 | XP_006230569.2 | 3 | 2542 | ||

Nlrp6
Genomic Map