siRNA Details
| Gene Symbol: UST4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134379.1 | NP_599206.1 | 2 | 554 | ||
| GenBank mRNA | AJ132859.1 | - | 2 | 554 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UST4r| Gene Symbol: UST4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134379.1 | NP_599206.1 | 2 | 554 | ||
| GenBank mRNA | AJ132859.1 | - | 2 | 554 | ||