siRNA Details
| Gene Symbol: Phldb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191578.1 | NP_001178507.1 | 6 | 1986 | ||
| XM_006242875.2 | XP_006242937.1 | 9 | 2459 | |||
| XM_006242881.2 | XP_006242943.1 | 9 | 2455 | |||
| XM_008766141.1 | XP_008764363.1 | 9 | 2456 | |||
| GenBank mRNA | X74226.1 | - | 2 | 467 | ||

Phldb1
Genomic Map