siRNA Details
| Gene Symbol: Crtac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134401.2 | NP_599228.2 | 11 | 1486 | ||
| XM_006231385.2 | XP_006231447.1 | 11 | 1548 | |||
| GenBank mRNA | BC089944.1 | - | 9 | 1017 | ||
| U78304.1 | - | 2 | 127 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crtac1| Gene Symbol: Crtac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134401.2 | NP_599228.2 | 11 | 1486 | ||
| XM_006231385.2 | XP_006231447.1 | 11 | 1548 | |||
| GenBank mRNA | BC089944.1 | - | 9 | 1017 | ||
| U78304.1 | - | 2 | 127 | |||