siRNA Details
| Gene Symbol: Zg16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134409.2 | NP_599236.2 | 3 | 139 | ||
| GenBank mRNA | AB095177.1 | - | 3 | 139 | ||
| CF249177.1 | - | 2 | 111 | |||
| Z30584.1 | - | 3 | 168 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zg16| Gene Symbol: Zg16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134409.2 | NP_599236.2 | 3 | 139 | ||
| GenBank mRNA | AB095177.1 | - | 3 | 139 | ||
| CF249177.1 | - | 2 | 111 | |||
| Z30584.1 | - | 3 | 168 | |||