siRNA Details
| Gene Symbol: LOC100910481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752277.3 | XP_003752325.1 | 185 | |||
| Gene Symbol: Marc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134410.2 | NP_599237.1 | 1 | 197 | ||
| GenBank mRNA | AF095741.1 | - | 1 | 198 | ||
| BC061734.1 | - | 1 | 224 | 5598768 | ||

LOC100910481
Genomic Map