siRNA Details
| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134417.1 | NP_599244.1 | 1, 2 | 251 | ||
| XM_006256345.2 | XP_006256407.1 | 1, 2 | 304 | |||
| GenBank mRNA | AY014898.1 | - | 1, 2 | 251 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ipmk| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134417.1 | NP_599244.1 | 1, 2 | 251 | ||
| XM_006256345.2 | XP_006256407.1 | 1, 2 | 304 | |||
| GenBank mRNA | AY014898.1 | - | 1, 2 | 251 | ||