siRNA Details
| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134461.1 | NP_604456.1 | 3, 4 | 410 | ||
| XM_006229729.2 | XP_006229791.2 | 3, 4 | 647 | |||
| XM_006229730.2 | XP_006229792.1 | 3, 4 | 539 | |||
| XM_006229731.2 | XP_006229793.1 | 3, 4 | 783 | |||
| GenBank mRNA | AF484958.1 | - | 3, 4 | 410 | ||
| FQ223532.1 | - | 3, 4 | 504 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007602, Y10656, AK164293, AK170034, BC014767, and others.

Capn5
Genomic Map