siRNA Details
| Gene Symbol: Atp2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134462.1 | NP_604457.1 | 1, 2 | 189 | ||
| GenBank mRNA | AF484685.1 | - | 1, 2 | 189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp2c2| Gene Symbol: Atp2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134462.1 | NP_604457.1 | 1, 2 | 189 | ||
| GenBank mRNA | AF484685.1 | - | 1, 2 | 189 | ||