siRNA Details
| Gene Symbol: Scp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138508.4 | NP_612517.2 | 2 | 111 | ||
| GenBank mRNA | BC081713.1 | - | 2 | 124 | 7109192 | |
| M57453.1 | - | 2 | 111 | |||
| M62763.1 | - | 2 | 138 | |||
| X60654.1 | - | 2 | 91 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scp2| Gene Symbol: Scp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138508.4 | NP_612517.2 | 2 | 111 | ||
| GenBank mRNA | BC081713.1 | - | 2 | 124 | 7109192 | |
| M57453.1 | - | 2 | 111 | |||
| M62763.1 | - | 2 | 138 | |||
| X60654.1 | - | 2 | 91 | |||