siRNA Details
| Gene Symbol: Pabpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134353.3 | NP_599180.1 | 3 | 621 | ||
| GenBank mRNA | AJ298278.1 | - | 3 | 621 | ||
| BC083176.1 | - | 3 | 865 | 7097075 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pabpc1| Gene Symbol: Pabpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134353.3 | NP_599180.1 | 3 | 621 | ||
| GenBank mRNA | AJ298278.1 | - | 3 | 621 | ||
| BC083176.1 | - | 3 | 865 | 7097075 | ||