siRNA Details
Gene Symbol: Slc12a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134363.1 | NP_599190.1 | 3 | 342 | ||
GenBank mRNA | EF641113.1 | - | 355 | |||
U55816.1 | - | 3 | 342 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc12a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134363.1 | NP_599190.1 | 3 | 342 | ||
GenBank mRNA | EF641113.1 | - | 355 | |||
U55816.1 | - | 3 | 342 |