siRNA Details
Gene Symbol: Spata19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134384.1 | NP_599211.1 | 3 | 197 | ||
GenBank mRNA | AB057408.1 | - | 3 | 197 | ||
BC081729.1 | - | 3 | 214 | 7132055 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spata19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134384.1 | NP_599211.1 | 3 | 197 | ||
GenBank mRNA | AB057408.1 | - | 3 | 197 | ||
BC081729.1 | - | 3 | 214 | 7132055 |