siRNA Details
| Gene Symbol: Pmfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134393.2 | NP_599220.1 | 3, 4 | 450 | ||
| XM_006255570.2 | XP_006255632.2 | 4, 5 | 1194 | |||
| XM_006255571.2 | XP_006255633.2 | 4, 5 | 1194 | |||
| XM_006255572.2 | XP_006255634.1 | 4, 5 | 1508 | |||
| XM_006255573.2 | XP_006255635.1 | 4, 5 | 749 | |||
| XM_006255574.2 | XP_006255636.2 | 4, 5 | 1192 | |||
| XM_006255576.2 | XP_006255638.2 | 4, 5 | 1193 | |||
| XM_008772468.1 | XP_008770690.1 | 3, 4 | 1038 | |||
| XM_008772469.1 | XP_008770691.1 | 3, 4 | 592 | |||
| XM_008772470.1 | XP_008770692.1 | 4, 5 | 1194 | |||
| GenBank mRNA | AF092090.2 | - | 3, 4 | 455 | ||

Pmfbp1
Genomic Map