siRNA Details
| Gene Symbol: Micu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134396.2 | NP_599223.2 | 4 | 409 | ||
| GenBank mRNA | AF327513.1 | - | 4 | 409 | ||
| DY317464.1 | - | 2 | 59 | |||
| EV765275.1 | - | 4 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Micu2| Gene Symbol: Micu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134396.2 | NP_599223.2 | 4 | 409 | ||
| GenBank mRNA | AF327513.1 | - | 4 | 409 | ||
| DY317464.1 | - | 2 | 59 | |||
| EV765275.1 | - | 4 | 416 | |||