siRNA Details
| Gene Symbol: Abtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134403.2 | NP_599230.2 | 9, 10 | 2396 | ||
| GenBank mRNA | AB000216.1 | - | 9, 10 | 2406 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abtb2| Gene Symbol: Abtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134403.2 | NP_599230.2 | 9, 10 | 2396 | ||
| GenBank mRNA | AB000216.1 | - | 9, 10 | 2406 | ||