siRNA Details
| Gene Symbol: Col5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134452.1 | NP_604447.1 | 3 | 307 | ||
| XM_006233873.2 | XP_006233935.1 | 3 | 713 | |||
| GenBank mRNA | AF272662.1 | - | 3 | 307 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col5a1| Gene Symbol: Col5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134452.1 | NP_604447.1 | 3 | 307 | ||
| XM_006233873.2 | XP_006233935.1 | 3 | 713 | |||
| GenBank mRNA | AF272662.1 | - | 3 | 307 | ||