siRNA Details
| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134461.1 | NP_604456.1 | 6 | 863 | ||
| XM_006229729.2 | XP_006229791.2 | 6 | 1100 | |||
| XM_006229730.2 | XP_006229792.1 | 6 | 992 | |||
| XM_006229731.2 | XP_006229793.1 | 6 | 1236 | |||
| XM_008759673.1 | XP_008757895.1 | 5 | 1027 | |||
| GenBank mRNA | AF484958.1 | - | 6 | 863 | ||
| FQ223532.1 | - | 6 | 957 | |||

Capn5
Genomic Map