siRNA Details
| Gene Symbol: Nme7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138532.1 | NP_612541.1 | 6 | 606 | ||
| XM_008769600.1 | XP_008767822.1 | 4 | 569 | |||
| XM_008769601.1 | XP_008767823.1 | 4 | 571 | |||
| GenBank mRNA | AF202049.1 | - | 6 | 606 | ||
| BC060314.1 | - | 6 | 627 | 6918381 | ||

Nme7
Genomic Map