siRNA Details
| Gene Symbol: Elovl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134382.1 | NP_599209.1 | 3 | 276 | ||
| XM_006243417.1 | XP_006243479.1 | 4 | 441 | |||
| GenBank mRNA | AB071985.1 | - | 3 | 276 | ||
| HQ404314.1 | - | 3 | 276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elovl5| Gene Symbol: Elovl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134382.1 | NP_599209.1 | 3 | 276 | ||
| XM_006243417.1 | XP_006243479.1 | 4 | 441 | |||
| GenBank mRNA | AB071985.1 | - | 3 | 276 | ||
| HQ404314.1 | - | 3 | 276 | |||