siRNA Details
| Gene Symbol: Spata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134392.2 | NP_599219.2 | 7 | 800 | ||
| GenBank mRNA | AF291466.1 | - | 7 | 1016 | ||
| BC126081.1 | - | 7 | 733 | |||
| BC158581.1 | - | 7 | 749 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata6| Gene Symbol: Spata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134392.2 | NP_599219.2 | 7 | 800 | ||
| GenBank mRNA | AF291466.1 | - | 7 | 1016 | ||
| BC126081.1 | - | 7 | 733 | |||
| BC158581.1 | - | 7 | 749 | |||