siRNA Details
| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134398.2 | NP_599225.1 | 7, 8 | 741 | ||
| XM_006243223.2 | XP_006243285.1 | 7, 8 | 770 | |||
| GenBank mRNA | AF178669.1 | - | 7, 8 | 743 | ||
| FQ224819.1 | - | 7, 8 | 713 | |||

Aagab
Genomic Map