siRNA Details
| Gene Symbol: Akr7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134407.1 | NP_599234.1 | 6 | 798 | ||
| GenBank mRNA | AB037424.1 | - | 6 | 798 | ||
| AF503514.1 | - | 6 | 836 | |||
| AJ271883.1 | - | 6 | 801 | |||
| BC061816.1 | - | 6 | 801 | 5598724 | ||
| FQ230711.1 | - | 3 | 207 | |||
| FQ231883.1 | - | 3 | 207 | |||

Akr7a2
Genomic Map