siRNA Details
| Gene Symbol: Lphn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF063102.1 | - | 24 | 4324 | ||
| AF081148.1 | - | 23 | 4665 | |||
| AF081149.1 | - | 22 | 4569 | |||
| AF081150.1 | - | 23 | 4698 | |||
| AF081151.1 | - | 24 | 4710 | |||
| AF081152.1 | - | 23 | 4614 | |||
| AF081153.1 | - | 24 | 4743 | |||
| CO556290.1 | - | 1 | 427 | |||

Lphn2
Genomic Map