siRNA Details
Gene Symbol: Cep170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB038495.1 | - | 5 | 1340 |
Gene Symbol: LOC100909664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249680.2 | XP_006249742.1 | 8 | 2565 | ||
XM_006249681.2 | XP_006249743.1 | 8 | 2565 |
