siRNA Details
| Gene Symbol: BFAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016561.2 | NP_057645.1 | 4 | 789 | ||
| XM_005255350.2 | XP_005255407.1 | 3 | 564 | |||
| GenBank mRNA | AF173003.1 | - | 4 | 661 | ||
| AK291114.1 | - | 4 | 597 | |||
| AK300778.1 | - | 2 | 362 | |||
| AK313890.1 | - | 3 | 597 | |||
| BC003054.1 | - | 4 | 620 | 3537783 | ||

BFAR
Genomic Map