siRNA Details
| Gene Symbol: Col5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134452.1 | NP_604447.1 | 65 | 5335 | ||
| XM_006233873.2 | XP_006233935.1 | 65 | 5741 | |||
| GenBank mRNA | AF272662.1 | - | 65 | 5335 | ||
| AJ005394.1 | - | 4 | 411 | |||
| FQ233333.1 | - | 5 | 675 | |||

Col5a1
Genomic Map