siRNA Details
| Gene Symbol: Cd99l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134459.1 | NP_604454.1 | 6 | 437 | ||
| XM_006251676.2 | XP_006251738.1 | 6 | 473 | |||
| XM_008770596.1 | XP_008768818.1 | 6 | 463 | |||
| GenBank mRNA | AB031014.1 | - | 6 | 437 | ||
| AF481858.1 | - | 6 | 384 | |||
| Gene Symbol: RGD1559441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237887.2 | XP_006237949.1 | 5 | 390 | ||
| XM_008763576.1 | XP_008761798.1 | 10 | 1158 | |||

Cd99l2
Genomic Map