siRNA Details
| Gene Symbol: Adra2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138505.2 | NP_612514.2 | 1 | 1297 | ||
| GenBank mRNA | M32061.1 | - | 1 | 1294 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adra2b| Gene Symbol: Adra2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138505.2 | NP_612514.2 | 1 | 1297 | ||
| GenBank mRNA | M32061.1 | - | 1 | 1294 | ||
Set Membership: |
|