siRNA Details
| Gene Symbol: Dkk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138519.2 | NP_612528.2 | 4, 5 | 570 | ||
| XM_006230009.2 | XP_006230071.1 | 4, 5 | 578 | |||
| GenBank mRNA | AF245040.1 | - | 3, 4 | 436 | ||
| BC085702.1 | - | 3, 4 | 430 | |||
| BC160821.1 | - | 4, 5 | 570 | 7321198 | ||

Dkk3
Genomic Map