siRNA Details
| Gene Symbol: Stambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138531.2 | NP_612540.1 | 9, 10 | 1350 | ||
| XM_006236726.2 | XP_006236788.1 | 10, 11 | 2017 | |||
| XM_006236727.2 | XP_006236789.1 | 11, 12 | 2135 | |||
| XM_008763045.1 | XP_008761267.1 | 9, 10 | 3678 | |||
| XM_008763046.1 | XP_008761268.1 | 10, 11 | 2698 | |||
| XM_008763047.1 | XP_008761269.1 | 10, 11 | 3115 | |||
| XM_008763048.1 | XP_008761270.1 | 9, 10 | 2997 | |||
| GenBank mRNA | AY083159.1 | - | 8, 9 | 1174 | ||
| BC061711.1 | - | 9, 10 | 1350 | 5597943 | ||

Stambp
Genomic Map