siRNA Details
| Gene Symbol: Tgoln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138840.2 | NP_620195.1 | 2 | 171 | ||
| GenBank mRNA | BC098512.1 | - | 2 | 171 | 7382894 | |
| X53565.1 | - | 2 | 169 | |||
| X64600.1 | - | 2 | 169 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgoln2| Gene Symbol: Tgoln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138840.2 | NP_620195.1 | 2 | 171 | ||
| GenBank mRNA | BC098512.1 | - | 2 | 171 | 7382894 | |
| X53565.1 | - | 2 | 169 | |||
| X64600.1 | - | 2 | 169 | |||