siRNA Details
| Gene Symbol: Cyb5r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138877.1 | NP_620232.1 | 2, 3 | 196 | ||
| XM_006242065.2 | XP_006242127.1 | 2, 3 | 224 | |||
| GenBank mRNA | D00636.1 | - | 2, 3 | 196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyb5r3| Gene Symbol: Cyb5r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138877.1 | NP_620232.1 | 2, 3 | 196 | ||
| XM_006242065.2 | XP_006242127.1 | 2, 3 | 224 | |||
| GenBank mRNA | D00636.1 | - | 2, 3 | 196 | ||