siRNA Details
| Gene Symbol: Sele | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138879.1 | NP_620234.1 | 2 | 199 | ||
| XM_008769608.1 | XP_008767830.1 | 3 | 581 | |||
| GenBank mRNA | L25527.1 | - | 2 | 199 | ||
| X69981.1 | - | 2 | 251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sele| Gene Symbol: Sele | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138879.1 | NP_620234.1 | 2 | 199 | ||
| XM_008769608.1 | XP_008767830.1 | 3 | 581 | |||
| GenBank mRNA | L25527.1 | - | 2 | 199 | ||
| X69981.1 | - | 2 | 251 | |||