siRNA Details
| Gene Symbol: Hspb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138887.1 | NP_620242.1 | 2 | 271 | ||
| GenBank mRNA | D29960.1 | - | 2 | 271 | ||
| FQ214730.1 | - | 2 | 296 | |||
| FQ215336.1 | - | 2 | 300 | |||
| FQ217608.1 | - | 2 | 300 | |||
| FQ224974.1 | - | 2 | 300 | |||

Hspb6
Genomic Map