siRNA Details
Gene Symbol: Ehd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138890.2 | NP_620245.2 | 1 | 557 | ||
XM_006239774.2 | XP_006239836.1 | 1 | 691 | |||
GenBank mRNA | AF494093.2 | - | 1 | 557 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ehd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138890.2 | NP_620245.2 | 1 | 557 | ||
XM_006239774.2 | XP_006239836.1 | 1 | 691 | |||
GenBank mRNA | AF494093.2 | - | 1 | 557 |